Stampa…

2,3,4-Tri-O-isobutyryl-1-O-trichloroacetimidoyl-α-D-glucopyranuronic acid methyl ester ≥98%

Fornitore: Apollo Scientific
Attenzione

BICL4368-100MG BICL4368-250MG BICL4368-500MG
APOSBICL4368-100MGEA 364 EUR
APOSBICL4368-100MG APOSBICL4368-250MG APOSBICL4368-500MG
2,3,4-Tri-O-isobutyryl-1-O-trichloroacetimidoyl-α-D-glucopyranuronic acid methyl ester ≥98%
2,3,4-Tri-O-isobutyryl-1-O-trichloroacetimidoyl-α-D-glucopyranuronic acid methyl ester
Formula: C₂₁H₃₀Cl₃NO₁₀
Punto di fusione: 82…84 °C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD09817649
CAS Number: 150607-96-8

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR