Stampa…

4-Methylumbelliferyl sulphate potassium salt

Fornitore: Apollo Scientific
Attenzione

BIB6119-1G BIB6119-5G
APOSBIB6119-1GEA 79.9 EUR
APOSBIB6119-1G APOSBIB6119-5G
4-Methylumbelliferyl sulphate potassium salt
4-Methylumbelliferyl sulphate potassium salt
Formula: C₁₀H₇KO₆S
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00016970
CAS Number: 15220-11-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR