Stampa…
Surgical gloves, latex, sterile, Gammex® Latex
Fornitore: ANSELL MEDICAL PRODUCTS
330048060 330048055 330048070 330048075 330048080 330048065 330048085 330048090 330048095
ANSM330048060EA 82.7 EUR
ANSM330048060 ANSM330048055 ANSM330048070 ANSM330048075 ANSM330048080 ANSM330048065 ANSM330048085 ANSM330048090 ANSM330048095
Surgical gloves, latex, sterile, Gammex® Latex
Guanti
The Gammex® Latex glove is easily donned, even with damp hands. The micro-rough textured outer surface ensures a secure grip in wet or dry conditions to facilitate the most delicate instrument handling.
  • Sterile, powder-free, micro-textured surface
  • Comfortable, no glove retraction, even after long periods of use
  • Water-based synthetic (polyurethane) inner-coating facilitates double donning
  • Contains low levels of proteins, chemical additives, endotoxins and pyrogenics
  • Formulation is guaranteed free of thiurams and MBT
  • Adhesive band at the top of the straight