Stampa…
Hydrometer set
Fornitore: Amarell
H809630
AMARH809630EA 0 EUR
AMARH809630
Hydrometer set
Idrometri
Hydrometers with scale paper, colour code and type designation, weighted body, with (and without) thermometer inside the hydrometer body, round capillary, mercury filling suitable for government verification.
Informazioni per l'ordine: Supplied in a transparent plastic case.