Stampa…
TapeStation Systems Parts and Accessories
Fornitore: AGILENT
5067-5598 5067-5599 5067-5783 5067-5786 5188-8047 G2991-40007 G2992-40042 G2992-40046
732-3993EA 44.5 EUR
732-3993 732-3994 AGLS5067-5783 AGLS5067-5786 AGLS5188-8047 AGLSG2991-40007 AGLSG2992-40042 AGLSG2992-40046
TapeStation Systems Parts and Accessories
Sistemi per elettroforesi Sistemi per elettroforesi capillare
Accessories and spare parts for the 4150 and 4200 TapeStation systems like plates & foil seals, loading tips, TapeStation test tape, needle cartridge.
  • 8-well PCR tube strips or 96-well sample plates are available depending on sample throughput, bringing added flexibility
  • Needle cartridges enable fast and simple needle exchange to ensure proper maintenance of the TapeStation systems
  • TapeStation test tape is available for troubleshooting and running system diagnostics tests, the tapes are reusable and expire after two years
  • Loading tips are pre-packed