Stampa…
Storage and Assay Microplates
Fornitore: AGILENT
203426-100 201240-100 204600-100 204357-100
AGLS203426-100EA 179 EUR
AGLS203426-100 AGLS201240-100 AGLS204600-100 AGLS204357-100
Storage and Assay Microplates
Micropiastre Micropiastre Deep Well
For biological assays, low storage temperatures, and filtrate collection applications.
  • Compatible with all major automation platforms and benchtop devices
  • No risk feasibility testing – test a sample in your application
  • Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
  • Molded with chemical resistant polymers, for maintained performance with prolonged exposure to aggressive solutions and solvents
  • Sealable with heat and adhesive seals, or cap mats, for superior performance
  • Low temperature tolerant for refrigerator or freezer