Stampa…
Platinum Standard Solution, 1000 mg/L Pt in Diluted Hydrochloric Acid for ICP-MS
Fornitore: ACTIVE MOTIF
M244.2CP.L1
ACSDM244.2CP.L1EA 418 EUR
ACSDM244.2CP.L1
Platinum Standard Solution, 1000 mg/L Pt in Diluted Hydrochloric Acid for ICP-MS
Standard Standard cationici Standard di platino