Stampa…
Calcium Standard Solution, 10000 mg/L Ca in dil. Nitric Acid Standard for ICP-MS
Fornitore: ACTIVE MOTIF
M109.2NP.L1
ACSDM109.2NP.L1EA 231 EUR
ACSDM109.2NP.L1
Calcium Standard Solution, 10000 mg/L Ca in dil. Nitric Acid Standard for ICP-MS
Standard Standard cationici Standard di calcio
Ca 10000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Certificazioni: ISO 17034, ISO 17025, ISO 9001.