Stampa…

Ferric chloride solution_SC Reag. Ph. Eur. R1

Fornitore: ACTIVE MOTIF

Synonyms: Iron(III) chloride solution

1037801
ACSD1037801EA 165 EUR
ACSD1037801
Ferric chloride solution_SC Reag. Ph. Eur. R1
Iron(III) chloride solutions
Formula: FeCl₃·6H₂O
Peso molecolare: 270.3 g/mol
MDL Number: MFCD00149712
CAS Number: 10025-77-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR