Stampa…

Clemastine fumarate

Fornitore: Thermo Fisher Scientific

465420010 465422500
ACRO465420010EA 615 EUR
ACRO465420010 ACRO465422500
Clemastine fumarate
Clemastine fumarate

  • Bioactive

Formula: C₂₅H₃₀ClNO₅
Punto di fusione: 158…162 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00137486
CAS Number: 14976-57-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %
Specific optical rotation +15° to +18° (20°C, 589 nm) (10 mg/ml methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR