Stampa…

Eplerenone

Fornitore: Thermo Fisher Scientific

Synonyms: 9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-pregn-4-ene-7alpha,21-dicarboxylic Acid gamma-Lactone Methyl Ester

462970010 462972500
ACRO462970010EA 650 EUR
ACRO462970010 ACRO462972500
Eplerenone
Eplerenone

  • Bioactive

Formula: C₂₄H₃₀O₆
Punto di fusione: 242 °C (dec.)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD07783647
CAS Number: 107724-20-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR