Stampa…

(R)-α-Methyltryptophan hemihydrate 98%, 98% ee

Fornitore: Thermo Fisher Scientific

441191000
ACRO441191000EA 183 EUR
ACRO441191000
(R)-α-Methyltryptophan hemihydrate 98%, 98% ee
(R)-alpha-Methyltryptophan hemihydrate
Formula: C₁₂H₁₄N₂O₂·0.5H₂O
Punto di fusione: >231 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD19687046
CAS Number: 56452-52-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Enantiomeric excess ≥98.0 %
Specific optical rotation +14° to +18° (20°C, 589 nm) (c=0.9 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR