Stampa…

Orange II, puro, certificato

Fornitore: Thermo Fisher Scientific
Pericolo

416560250 416561000
ACRO416560250EA 26.2 EUR
ACRO416560250 ACRO416561000
Orange II, puro, certificato
Orange II

CAS No.: 633-96-5

Formula: C₁₆H₁₁N₂NaO₄S
Punto di fusione: 164 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011657
CAS Number: 633-96-5

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to dark orange-red Powder
Infrared spectrum Conforms
Dye content ≥85 %
UV Lambda max 480 to 486 nm (c=0.01 g/l, H2O)
Absorption Ratio P-15/P+15 0.93 to 0.98
BSC Certification Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR