Stampa…

Dodecyltrimethylammonium bromide 99%, puro

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide

409311000 409310250 409315000
ACRO409311000EA 407 EUR
ACRO409311000 ACRO409310250 ACRO409315000
Dodecyltrimethylammonium bromide 99%, puro
Dodecyltrimethylammonium bromide
Formula: C₁₅H₃₄BrN
Punto di fusione: 246 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011767
CAS Number: 1119-94-4
EINECS: 214-290-3
UN: 3077
ADR: 9,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Solubility (10 % in water) Clear to slightly hazy
Color of solution ≤100 APHA (10% aq. soln.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR