Stampa…

Sodium decyl sulphate per HPLC

Fornitore: Thermo Fisher Scientific
Pericolo

406220050 406220250 406221000
ACRO406220050EA 168 EUR
ACRO406220050 ACRO406220250 ACRO406221000
Sodium decyl sulphate per HPLC
Sodium decyl sulphate
Formula: C₁₀H₂₁NaO₄S
Punto di fusione: 191,85 °C
Densità: 1,207 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00041881
CAS Number: 142-87-0
EINECS: 205-568-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤1.5 % (K.F.)
Solubility (25 % in water at 25°C) Clear colorless to light yellow
UV at 280 nm A: ≤0.01
Titration after Ion exchange ≥98.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR