Stampa…

Sodium trifluoromethanesulphonate 98%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: Trifluoromethanesulphonic acid sodium salt

358750050 358750250
ACRO358750050EA 55.2 EUR
ACRO358750050 ACRO358750250
Sodium trifluoromethanesulphonate 98%
Sodium trifluoromethanesulphonate
Formula: CF₃NaO₃S
Peso molecolare: 172.06 g/mol
Punto di fusione: 253…255 °C
MDL Number: MFCD00061607
CAS Number: 2926-30-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to off-white Powder
Infrared spectrum Conforms
Assay ≥97.5 % (ex Na)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR