Stampa…

Hemin (from porcine) ≥99%

Fornitore: Thermo Fisher Scientific
Pericolo

345960010 345960050 345960250
ACRO345960010EA 50.7 EUR
ACRO345960010 ACRO345960050 ACRO345960250
Hemin (from porcine) ≥99%
Hemin chloride

Hemin derived from porcine is an iron-containing prosthetic group present in some proteins and can be an alternative source of iron within the host that contains a porphyrin ring containing a Fe2+ ion.
Hemin is a protoporphyrin IX containing a ferric iron (Fe3+) ion with a coordinating chloride ligand


  • It is used in cellular protection and controls biological mechanisms
  • It can stimulate globulin synthesis
  • It can be applied to study bacterial strains and growth conditions in the short-term colony assays and cell transfections
  • It is used during the preparation of agar medium for bacteria culture


This compound has been used in L929 and J774 murine macrophage cells in order to study conjugation, characterization and toxicity of lipophosphoglycan-polyacrylic acid conjugate for vaccination against leishmaniasis


Attenzione: RUO – Research Use Only

Formula: C₃₄H₃₂ClFeN₄O₄
MDL Number: MFCD00010726
CAS Number: 16009-13-5
UN: 3400
ADR: 4.2,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR