Stampa…
Chloroform : isoamyl alcohol (24:1) (v/v) per biologia molecolare DNase-, RNase-, protease-free
Fornitore: Thermo Fisher Scientific
Pericolo
327155000
ACRO327155000EA 206 EUR
ACRO327155000
Chloroform : isoamyl alcohol (24:1) (v/v) per biologia molecolare DNase-, RNase-, protease-free
Chloroform : isoamyl alcohol
Formula: CHCl₃,C₅H₁₂O
Punto di fusione: –63 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00171265
CAS Number: 67-66-3
UN: 1888
ADR: 6.1,III
Visualizza i codici
Appearance Clear solution
GC ≥99.0 % (isoamyl alcohol component)
Dnase,Rnase,Protease activity none detected