Stampa…

N-(p-Toluenesulphonyl)-L-phenylalanylchloromethylketone 99+%

Fornitore: Thermo Fisher Scientific
Pericolo

227800010 227800050
ACRO227800010EA 153 EUR
ACRO227800010 ACRO227800050
N-(p-Toluenesulphonyl)-L-phenylalanylchloromethylketone 99+%
N-(p-Toluenesulphonyl)-L-phenylalanylchloromethylketone

CAS No.: 402-71-1

Formula: C₁₇H₁₈ClNO₃S
Punto di fusione: 105…109 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00000935
CAS Number: 402-71-1

Visualizza i codici

Risultati del test di specificazione

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 105 to 109 °C
TLC ≥98.0 %
Specific optical rotation -86° to -88° (20 °C, 589 nm) (c=1, abs. ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR