Stampa…

Potassium-4-nitrophenyl sulphate 99+%

Fornitore: Thermo Fisher Scientific

Synonyms: 4-Nitrophenyl sulphate, potassium salt, Potassium 4-Nitrophenyl sulphate, 4-Nitrophenyl sulphate potassium salt, sulphuric acid 4-Nitrophenyl ester potassium salt

227490010 227490050
ACRO227490010EA 162 EUR
ACRO227490010 ACRO227490050
Potassium-4-nitrophenyl sulphate 99+%
Potassium-4-nitrophenyl sulphate

CAS No.: 6217-68-1

Formula: C₆H₄KNO₆S
Peso molecolare: 257.257 g/mol
Punto di fusione: 246…250 °C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00007466
CAS Number: 6217-68-1

Visualizza i codici

Risultati del test di specificazione

Appearance Yellow to khaki to brown Powder
Infrared spectrum Conforms
HPLC ≤0.05 % free p-nitrophenol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR