Stampa…

Cobalto (II) nitrato esaidrato 99+% per analisi

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate

219211000 219215000 219210025
ACRO219211000EA 93.7 EUR
ACRO219211000 ACRO219215000 ACRO219210025
Cobalto (II) nitrato esaidrato 99+% per analisi
Cobalto (II) nitrato esaidrato
Formula: Co(NO₃)₂·6H₂O
Punto di fusione: 55 °C
Densità: 1,88 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,II
Merck Index: 13,02469

Visualizza i codici

Risultati del test di specificazione

Appearance Pink to red to brown Adhering crystalline powder or crystals or chips
Titration Complexometric ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤20 ppm
Nickel (Ni) ≤500 ppm
Zinc (Zn) ≤50 ppm
Heavy metals (as Cu) ≤10 ppm
Insoluble matter ≤0.01 %

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR