Stampa…

Calcio acetato x-idrato 99%, extra puro

Fornitore: Thermo Fisher Scientific

211050010 211052500 211050051
ACRO211050010EA 72.8 EUR
ACRO211050010 ACRO211052500 ACRO211050051
Calcio acetato x-idrato 99%, extra puro
Calcio acetato x-idrato
Formula: (H₃CCOO)₂Ca·nH₂O
Densità: 1,5 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00150019
CAS Number: 114460-21-8
EINECS: 200-540-9

Visualizza i codici

Risultati del test di specificazione

Appearance White Powder and/or agglomerates
Infrared spectrum Conforms
Titration Complexometric 98.5 to 101.5 % (on anhydrous substance)
Water ≤7.0 % (K.F.)
pH 7.0 to 9.0 (10 % at 20°C)
Chloride (Cl) ≤300 ppm
Fluoride (F) ≤50 ppm
Sulfate (SO4) ≤500 ppm
Arsenic (As) ≤2 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR