Stampa…

Acid Red 18 (Food Red No. 102), puro

Fornitore: Thermo Fisher Scientific

Synonyms: Cochineal Red A, New Coccine, Scarlet 3R, Sodium 7-hydroxy-8-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1,3-disulphonate, Ponceau 4R, Cochenille red

190230250 190231000
ACRO190230250EA 43.8 EUR
ACRO190230250 ACRO190231000
Acid Red 18 (Food Red No. 102), puro
Acid Red 18 (Food Red No. 102)

CAS No.: 2611-82-7

Formula: C₂₀H₁₄N₂O₁₀S₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004084
CAS Number: 2611-82-7

Visualizza i codici

Risultati del test di specificazione

Appearance Red to brown Powder
Infrared spectrum Conforms
UV spectrum Complies
Solubility (0.1 % in water) Clear red to very dark red
Dye content ≥75 %
Insoluble matter ≤0.2 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR