Stampa…

Polivinil alcool, M.W. ∼ 85,000-124,000 95.5-96.5 %, hydrolysed

Fornitore: Thermo Fisher Scientific

Synonyms: PVOH, PVA, Mowiol®

183390010 183390025 183391000
ACRO183390010EA 171 EUR
ACRO183390010 ACRO183390025 ACRO183391000
Polivinil alcool, M.W. ∼ 85,000-124,000 95.5-96.5 %, hydrolysed
Polivinil alcool
Formula: [-CH₂CH(OH)-]n
Punto di fusione: 230…240 °C
Densità: 1,3 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to cream Granular powder
Infrared spectrum Conforms
Viscosity 27 to 31 cP (4 % in water)
Degree of hydrolysis 95.5 to 96.5 mol %
Ash ≤0.9 % (as Na2O)
pH 4.5 to 6.5 (4 % in water)
Volatile matter ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR