Stampa…

Olio silicone, 475 -525 mm²/s (25°C) for melting point and boiling point apparatuses

Fornitore: Thermo Fisher Scientific

Synonyms: Polyphenyl-methylsiloxane

163850010 163855000 163850025
ACRO163850010EA 197 EUR
ACRO163850010 ACRO163855000 ACRO163850025
Olio silicone, 475 -525 mm²/s (25°C) for melting point and boiling point apparatuses
Olio silicone
Punto di fusione: –50 °C
Densità: 0.96
Flash Pt: 300 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00132673
CAS Number: 68083-14-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR