Stampa…

(S)-(+)-Carvone (naturale) 96%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: D-p-Mentha-1(6),8-dien-2-one, D(+)-Carvone, (S)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one

150680250 150681000
ACRO150680250EA 32.3 EUR
ACRO150680250 ACRO150681000
(S)-(+)-Carvone (naturale) 96%
(S)-(+)-Carvone
Formula: C₁₀H₁₄O
Peso molecolare: 150.22 g/mol
Punto di ebollizione: 230
Densità: 0.956 g/cm³
Flash Pt: 97 °C
MDL Number: MFCD00062997
CAS Number: 2244-16-8
EINECS: 218-827-2

Visualizza i codici

Risultati del test di specificazione

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4960 to 1.5010 (20°C, 589 nm)
GC ≥95 %
Specific gravity (25°C) 0.953 to 0.962
Specific optical rotation +52.0° to +62.0° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR