Stampa…

3-Iodo-L-tyrosine 97%

Fornitore: Thermo Fisher Scientific

Synonyms: H-Tyr(3-I)-OH, (S)-2-Amino-3-(4-hydroxy-3-iodophenyl)propanoic acid

122440010 122440050 122440250
ACRO122440010EA 43 EUR
ACRO122440010 ACRO122440050 ACRO122440250
3-Iodo-L-tyrosine 97%
3-Iodo-L-tyrosine
Formula: C₉H₁₀INO₃
Peso molecolare: 307.1 g/mol
Punto di fusione: 196…198 °C (dec.)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00002608
CAS Number: 70-78-0
EINECS: 200-744-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤4 % (K.F.)
Solubility (5 % in 1 N HCl) Clear colorless to light yellow
HPLC ≥96.0 %
Specific optical rotation -3° to -5° (20°C, 589 nm) (c=5 in 1 N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR