Stampa…

Anti-Blood Group Lewis a Mouse Monoclonal Antibody [clone: 7LE]

Fornitore: Abcam
AB3967-100
ABCAAB3967-100EA 930 EUR
ABCAAB3967-100
Anti-Blood Group Lewis a Mouse Monoclonal Antibody [clone: 7LE]
Anticorpi
Attenzione:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production

More Product Information
Visualizza i codici

Specifiche

Environmentally Preferable
Tipo di anticorpi Blood Group Lewis A
Tipo anticorpo Primary
Clonalità Monoclonale
Coniugazione Non coniugato
Clone 7LE
Reattività Human,Mouse
Ospite Mouse
Isotipo IgG1
Western blot Yes
ELISA Yes
ImmunoChimica Yes
ImmunoFluorescenza Yes
Citometria a flusso Yes
Temperatura di conservazione Cool
Temperatura di spedizione Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR