Stampa…
Cy3.5 tetrazine
Fornitore: AAT BIOQUEST
903
AATB903EA 309 EUR
AATB903
Cy3.5 tetrazine
Cy3.5 tetrazine
Cy3.5 tetrazine is an excellent building block that can be readily used to label TCO-modified biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
  • Stable for 12 months
Quality tested in-house
Attenzione: For Research Use Only