Stampa…
XFD488 TCO
Fornitore: AAT BIOQUEST
4601
AATB4601EA 277 EUR
AATB4601
XFD488 TCO
XFD488 TCO
XFD488 contains the same fluorophore of Alexa Fluor® 488.
  • Stable for 12 months
Quality tested in-house
Attenzione: For Research Use Only