Stampa…

Biotin hydrazide, ReadiView™ biodetection

Fornitore: AAT BIOQUEST
Attenzione

Synonyms: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide

3055
AATB3055EA 307 EUR
AATB3055
Biotin hydrazide, ReadiView™ biodetection
Biotin hydrazide

Biotin/avidin complexes are widely applied for a variety of biological detections.


  • High fluorescence enhancement
  • Sensitive detection
  • Multiple applications

Formula: C₁₀H₁₈N₄O₂S
MDL Number: MFCD00078532
CAS Number: 66640-86-6
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4b827b6458d2ce2b42d75f39981279c84dcd306&vid=3dadffc3f3c2b1855a74bd95a7b767e3c5be148f&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR