Stampa…
MycoLight™ vPCR350
Fornitore: AAT BIOQUEST
24210
AATB24210EA 198 EUR
AATB24210
MycoLight™ vPCR350
MycoLight™ vPCR350
MycoLight™ vPCR350 is a novel non-fluorescent DNA modifier specifically designed for viability PCR (vPCR) targeting microorganisms such as bacteria, eukaryotes, viruses, and fungi.
  • Stable for 12 months
Quality tested in-house
Attenzione: For Research Use Only