Stampa…

Propidium iodide 10 mM in soluzione acquosa ≥95%

Fornitore: AAT BIOQUEST

17517
AATB17517EA 152 EUR
AATB17517
Propidium iodide 10 mM in soluzione acquosa ≥95%
Propidium iodide

Propidium iodide (PI) belongs to the same chemical class of ethidium bromide.


  • Intercalating Nucleic Acid Probe
  • Useful As Dead Cell Counterstain


Attenzione: For research use only.

Formula: C₂₇H₃₄I₂N₄
MDL Number: MFCD00011921
CAS Number: 25535-16-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR