Stampa…
Amplite® Colorimetric Fructose-6-Phosphate Activity Assay Kit
Fornitore: AAT BIOQUEST
11330
AATB11330EA 673 EUR
AATB11330
Amplite® Colorimetric Fructose-6-Phosphate Activity Assay Kit
Saggi Saggi enzimatici
Designed for consistent and dependable results. The Amplite® colorimetric fructose-6-phosphate assay kit is a highly sensitive and simple method of quantifying F6P present in a variety of samples.
  • Robust protocol
  • Absorbance measured at 450 nm
  • Six-month shelf life
One unit (U) is the amount of enzyme that catalyses the reaction of 1 µmol of substrate per minute.
Informazioni per l'ordine: Shipped with blue ice.
Attenzione: Research use only.