Stampa…

Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 860)

Fornitore: AAT BIOQUEST
101920R0 101920R1
AATB101920R0EA 371 EUR
AATB101920R0 AATB101920R1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 860)
Anticorpi
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attenzione: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: iFluor® 860
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR