Stampa…
Decontamination solution for autoclaves Autoclave-ExitusPlus™
Fornitore: PanReac AppliChem
A7600.1000
A7600.1000EA 85.3 EUR
A7600.1000
Decontamination solution for autoclaves Autoclave-ExitusPlus™
Soluzioni detergenti Decontaminanti
Autoclaving is badly suited for the sustained removal of nucleic acids. DNA molecules in recombinant microorganisms are merely fragmented in the autoclave and subsequently released with the vapours when opening. By adding Autoclave-ExitusPlus™ to residual cultures or buffer solutions, the nucleic acids are effectively destroyed during autoclaving.