Stampa…

Triethylammonium bromide 98%

Fornitore: Thermo Fisher Scientific
Attenzione

A17926.22 A17926.36
A17926.22EA 39.4 EUR
A17926.22 A17926.36
Triethylammonium bromide 98%
Triethylammonium bromide

Triethylamine hydrobromide is used to prepare dimethyl -alfa-diethylaminomaleate by reacting with dimethyl acetylenedicarbosylate. It is involved in the preparation of triphenylpropargylphosphonium bromide.


Purity: 98% CAS No.: 636-70-4 Molecular Formula: C16H15N.HBr Formula Weight: 182.10 Odour: Amine-like Melting Point: ca 250° dec. MDL No.: MFCD00054287

Formula: C₆H₁₅N·HBr
Punto di fusione: ∼ 250 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00054287
CAS Number: 636-70-4
EINECS: 211-263-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR