Stampa…

Tetramethylphosphonium bromide ≥98%

Fornitore: Thermo Fisher Scientific
Attenzione

A17907.03 A17907.06
A17907.03EA 65.4 EUR
A17907.03 A17907.06
Tetramethylphosphonium bromide ≥98%
Tetramethylphosphonium bromide

Purity: 98% CAS No.: 4519-28-2 Molecular Formula: (CH3)4PBr Formula Weight: 171.03 MDL No.: MFCD00011802

Formula: C₄H₁₂BrP
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011802
CAS Number: 4519-28-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR