Stampa…

(Ethyl)triphenylphosphonium iodide ≥98%

Fornitore: Thermo Fisher Scientific
Pericolo

A17533.14 A17533.22 A17533.36
A17533.14EA 32.3 EUR
A17533.14 A17533.22 A17533.36
(Ethyl)triphenylphosphonium iodide ≥98%
(Ethyl)triphenylphosphonium iodide

Purity: 98+% CAS No.: 4736-60-1 Molecular Formula: CH3CH2P(C6H5)3I Formula Weight: 418.26 Melting Point: 164-169° Beilstein Reference: 3659323 UN No.: 2811 MDL No.: MFCD00040352

Formula: C₂₀H₂₀IP
Punto di fusione: 164…169 °C
Densità: 1 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00040352
CAS Number: 4736-60-1
EINECS: 225-245-2
UN: 2811
ADR: 6.1,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR