Stampa…

1,2-Cyclohexanedione dioxime ≥97%

Fornitore: Thermo Fisher Scientific

A16788.06 A16788.14 A16788.22
A16788.06EA 75.7 EUR
A16788.06 A16788.14 A16788.22
1,2-Cyclohexanedione dioxime ≥97%
1,2-Cyclohexanedione dioxime

Purity: 97% CAS No.: 492-99-9 Molecular Formula: C6H10N2O2 Formula Weight: 142.16 Melting Point: 181-188° dec. Beilstein Reference: 1909047 Merck Reference: 14,6560 MDL No.: MFCD00001663

Formula: C₆H₁₀N₂O₂
Punto di fusione: 181…188 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00001663
CAS Number: 492-99-9
EINECS: 207-769-3
Merck Index: 14,6560

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR