Stampa…

Benzyl-4-chlorophenyl ketone ≥98%

Fornitore: Thermo Fisher Scientific

A15959.06 A15959.14 A15959.22
A15959.06EA 45.1 EUR
A15959.06 A15959.14 A15959.22
Benzyl-4-chlorophenyl ketone ≥98%
Benzyl-4-chlorophenyl ketone

Purity: 98% CAS No.: 1889-71-0 Molecular Formula: C14H11ClO Formula Weight: 230.69 Melting Point: 103-107° Beilstein Reference: 976443

Formula: C₁₄H₁₁ClO
Punto di fusione: 103…107 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016342
CAS Number: 1889-71-0

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR