Stampa…

Tetraphenylphosphonium bromide ≥98%

Fornitore: Thermo Fisher Scientific
Attenzione

A15860.09 A15860.18 A15860.22
A15860.09EA 37.4 EUR
A15860.09 A15860.18 A15860.22
Tetraphenylphosphonium bromide ≥98%
Tetraphenylphosphonium bromide

Purity: 98+% CAS No.: 2751-90-8 Molecular Formula: (C6H5)4PBr Formula Weight: 419.31 Melting Point: 296-302° Beilstein Reference: 3922383 Merck Reference: 14,9237 MDL No.: MFCD00011915

Formula: C₂₄H₂₀BrP
Punto di fusione: 296…302 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011915
CAS Number: 2751-90-8
EINECS: 220-393-4
Merck Index: 14,9237

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR