Stampa…

Allantoin ≥98%

Fornitore: Thermo Fisher Scientific

A15571.30 A15571.0B A15571.0I
A15571.30EA 51.3 EUR
A15571.30 A15571.0B A15571.0I
Allantoin ≥98%
Allantoin

Purity: 98% CAS No.: 97-59-6 Molecular Formula: C4H6N4O3 Formula Weight: 158.12 Melting Point: ca 230° dec. Beilstein Reference: 102364 Merck Reference: 14,258 MDL No.: MFCD00005260

Formula: C₄H₆N₄O₃
Punto di fusione: ∼ 230 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005260
CAS Number: 97-59-6
EINECS: 202-592-8
Merck Index: 14,258

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR