Stampa…

Tetraoctylammonium bromide ≥98%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: Tetra-n-octylammonium bromide

A14929.09 A14929.18 A14929.30
A14929.09EA 56.3 EUR
A14929.09 A14929.18 A14929.30
Tetraoctylammonium bromide ≥98%
Tetraoctylammonium bromide

Purity: 98+% CAS No.: 14866-33-2 Molecular Formula: [CH3(CH2)7]4NBr Formula Weight: 546.81 Melting Point: 94-99° Beilstein Reference: 4169176

Formula: C₃₂H₆₈BrN
Punto di fusione: 94…99 °C
Flash Pt: 100 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011863
CAS Number: 14866-33-2
EINECS: 238-936-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR