Stampa…

L-Threonine

Fornitore: PanReac AppliChem

Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

A1419.1000
A1419.1000EA 416 EUR
A1419.1000
L-Threonine
L-Threonine
Formula: C₄H₉NO₃
Peso molecolare: 119.12 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR