Stampa…

Phenylpyruvic acid sodium salt monohydrate 98%

Fornitore: Thermo Fisher Scientific

A14027.03 A14027.06 A14027.14 A14027.18
A14027.03EA 22.7 EUR
A14027.03 A14027.06 A14027.14 A14027.18
Phenylpyruvic acid sodium salt monohydrate 98%
Phenylpyruvic acid sodium salt monohydrate

Purity: 98% CAS No.: 122049-54-1 Molecular Formula: C9H7NaO3.H2O Formula Weight: 204.16 (186.14anhy) Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 4770978 MDL No.: MFCD00149109

Formula: C₉H₉NaO₄
Punto di fusione: >300 °C
MDL Number: MFCD00149109
CAS Number: 122049-54-1
EINECS: 204-053-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR