Stampa…

Titanium(IV) isopropoxide 95%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: Titanium tetraisopropanolate, Titanium (IV) isopropoxide

A13703.0E A13703.22 A13703.36
A13703.0EEA 309 EUR
A13703.0E A13703.22 A13703.36
Titanium(IV) isopropoxide 95%
Titanium(IV) isopropoxide

Purity: 95% CAS No.: 546-68-9 Molecular Formula: C12H28O4Ti Formula Weight: 284.23 Flash Point: 46°(115°F) Refractive Index: 1.4640 Melting Point: 16-20° Boiling Point: 232° Beilstein Reference: 3679474 Merck Reference: 14,9480 UN No.: 2413 MDL No.: MFCD00008871

Formula: C₁₂H₂₈O₄Ti
Punto di ebollizione: 232 °C
Punto di fusione: 16…20 °C
Densità: 0,955
Flash Pt: 46 °C (115 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00008871
CAS Number: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3,III
Merck Index: 14,9480

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR