Stampa…

Cyclopropyl methyl ketone ≥99%

Fornitore: Thermo Fisher Scientific
Pericolo

A13540.14 A13540.22 A13540.36
A13540.14EA 31.6 EUR
A13540.14 A13540.22 A13540.36
Cyclopropyl methyl ketone ≥99%
Cyclopropyl methyl ketone

Purity: 99% CAS No.: 765-43-5 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 16°(61°F) Refractive Index: 1.4240 Melting Point: -70° Boiling Point: 113-114° Beilstein Reference: 1633817 UN No.: 1992 MDL No.: MFCD00001297

Formula: C₅H₈O
Punto di ebollizione: 113…114 °C
Punto di fusione: < –70 °C
Densità: 0,898
Flash Pt: 16 °C (61 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00001297
CAS Number: 765-43-5
EINECS: 212-146-4
UN: 1992
ADR: 3,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR