Stampa…

Acido nicotinico (Niacin) ≥99%

Fornitore: Thermo Fisher Scientific
Attenzione

A12683.30 A12683.0B A12683.0I
A12683.30EA 35.1 EUR
A12683.30 A12683.0B A12683.0I
Acido nicotinico (Niacin) ≥99%
Acido nicotinico (Niacin)

Purity: 99% CAS No.: 59-67-6 Molecular Formula: C6H5NO2 Formula Weight: 123.11 Flash Point: 293°(559°F) Physical Form: Powder Density: 1.4 Appearance: White Odour: Odourless pH: 2.7 Melting Point: 236-239° Beilstein Reference: 109591 Merck Reference: 14,6525 MDL No.: MFCD00006391


Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP

Formula: C₆H₅NO₂
Punto di ebollizione: 258 °C (1013 hPa)
Punto di fusione: 236…239 °C
Densità: 1,4
Flash Pt: 293 °C (559 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 14,6525

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR