Stampa…

Piperidina 99

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Hexahydropyridine, Hexazane, Pentamethyleneimine

A12442.0F A12442.AE A12442.AP A12442.PI
A12442.0FEA 126 EUR
A12442.0F A12442.AE A12442.AP A12442.PI
Piperidina 99
Piperidina

Purity: 99% CAS No.: 110-89-4 Molecular Formula: C5H11N Formula Weight: 85.15 Flash Point: 3°(37°F) Physical Form: Liquid Density: 0.862 Refractive Index: 1.4520 Sensitivity: Air Sensitive Appearance: Colourless Melting Point: -11° Boiling Point: 105-106° Beilstein Reference: 102438 Merck Reference: 14,7468 UN No.: 2401 MDL No.: MFCD00005979

Formula: C₅H₁₁N
Peso molecolare: 85.15 g/mol
Punto di ebollizione: 105…106 °C
Punto di fusione: –11 °C
Densità: 0.862
Flash Pt: 3 °C (37 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005979
CAS Number: 110-89-4
EINECS: 203-813-0
UN: 2401
ADR: 8 I
Merck Index: 14,7468

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR